missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMFR Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 16675-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Autocrine motility factor is a tumor motility-stimulating protein secreted by tumor cells. The protein encoded by this gene is a glycosylated transmembrane protein and a receptor for autocrine motility factor. The receptor, which shows some sequence similarity to tumor protein p53, is localized to the leading and trailing edges of carcinoma cells.Especificaciones
AMFR | |
Polyclonal | |
Unconjugated | |
AMFR | |
AMF receptor, isoform 2, AMFR, GP78, RING finger protein 45, RNF45 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23802, 267, 361367 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9R049, Q9UKV5 | |
Amfr | |
AMFR Fusion Protein Ag10178 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |