Learn More
Cytiva Amersham™ Hyperfilm™ ECL
Product Code. 10534205 Shop All Cytiva Products
Change view
Click to view available options
Dimensions (LxW):
12.7 x 17.8 cm (5 x 7 in.)
18 x 24 cm (7.1 x 9.5 in.)
20.3 x 25.4 cm (8 x 10 in.)
Quantity:
100 Sheets/Box
50 Sheets/Box
Unit Size:
100 sheets
50 sheets
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (LxW) Quantity Unit Size
10534205 18 x 24 cm (7.1 x 9.5 in.) 100 Sheets/Box 100 sheets
10752067 12.7 x 17.8 cm (5 x 7 in.) 50 Sheets/Box 50 sheets
10479214 20.3 x 25.4 cm (8 x 10 in.) 50 Sheets/Box 50 sheets
10607665 18 x 24 cm (7.1 x 9.5 in.) 50 Sheets/Box 50 sheets
10075224 20.3 x 25.4 cm (8 x 10 in.) 100 Sheets/Box 100 sheets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10534205 Supplier Cytiva Supplier No. 28906837
  • ONLINE EXCLUSIVE PRICE
    545.71€
    480.73€ / 100 hojas
    Ahorro 64.98€
    11% Descuento
 Restricted Item
En stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy
Use with all blue and green light chemiluminescent systems. Cytiva Amersham™ Hyperfilm™ ECL is constructed from a clear base coated on both sides with a photographic emulsion that is protected by an anti-scratch layer.
  • Antistatic coating reduces risk of visible artifacts
  • Clear base offers high contrast for easy reading of images
  • Suitable for use with all blue and green light chemiluminescent systems
  • Film can be processed in automatic processors or manually using most common X-ray film developers and fixers
  • Chemiluminescent detection in Western, Southern, and Northern blots
TRUSTED_SUSTAINABILITY
Specifications
Product Type Amersham Hyperfilm ECL
For Use With (Application) Chemiluminescent detection in Western, Southern, and Northern blots.
Quantity 100 Sheets/Box
Content And Storage 4deg;C
Dimensions (LxW) 18 x 24 cm (7.1 x 9.5 in.)