missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RNAqueous-Midi Total RNA Isolation Kit

RNA isolation for midrange amounts of starting material

340.00€ - 345.00€

Especificaciones

Para utilizar con (aplicación) Northern Blotting, Nuclease Protection Assays, RT-PCR (Endpoint), Real-Time PCR, cDNA Library Construction
Cantidad. 15 preps
Tipo de muestra Tissue Cells
Objetivo Total RNA
Ver más especificaciones

Productos 2
Código de producto Marca Tipo de producto Incluye Precio Cantidad y disponibilidad  
Código de producto Marca Tipo de producto Incluye Precio Cantidad y disponibilidad  
10015954 Invitrogen™
AM1911
Kit Formaldehyde Load Dye, Lysis/Binding Solution, Water for 64% Ethanol, Lithium, Chloride Solution, Wash Solution No. 1 and Wash Solution 2/3 Concentrate, Syringe Filter Units, Collection Tubes and Elution Solution 340.00€
cada uno Ver productos alternativos
Ahorre hasta 
»
Fecha de envío estimada
del proveedor 05-06-2019
Añadir a la cesta
 
10187674 Invitrogen™
AM1911M
Kit with manual Formaldehyde Load Dye, Lysis/Binding Solution, Water for 64% Ethanol, Lithium, Chloride Solution, Wash Solution No. 1 and Wash Solution 2/3 Concentrate, Syringe Filter Units, Collection Tubes and Elution Solution, Manual 345.00€
cada uno Ver productos alternativos
Ahorre hasta 
»
Fecha de envío estimada
del proveedor 05-06-2019
Añadir a la cesta
 
Descripción

Descripción

The Ambion RNAqueous-Midi Kit is for phenol-free total RNA isolation from 0.1g to 0.5g of tissue or 107 to 108 cells using a guanidinium-based lysis/;denaturant and glass fiber filter separation technology. The RNAqueous-Midi Kit provides enough reagents for 15 RNA purifications. The Ambion RNAqueous-Midi Kit uses a powerful chaotropic solution for cell lysis, followed by RNA binding to glass fiber housed in a luer-lock syringe filter. Washing and elution solutions are simply pushed through the filter using a standard syringe (not included). Vacuum filtration is not required; however, the filter units are compatible with standard vacuum manifolds for processing several samples at once.

  • Convenient syringe-based filtration
  • Compatible with vacuum manifolds when processing multiple samples

Not high-throughput compatible (Manual)

Applications:

DNA and RNA Purification and Analysis, RNA Extraction, Total RNA Isolation, Total RNA from Animal Cells and Tissues

Order Info

Shipping Condition: Room temperature

Especificaciones

Especificaciones

Northern Blotting, Nuclease Protection Assays, RT-PCR (Endpoint), Real-Time PCR, cDNA Library Construction
15 preps
Tissue Cells
Total RNA

For Research Use Only. Not for use in diagnostic procedures.