Learn More
Ambion™ Ambion™ In Vivo Pre-Designed siRNA
Ambion™ In Vivo Pre-Designed siRNA
Marca:  Ambion™ 4457310
Detalles adicionales : Peso : 0.01000kg
 Ver más versiones de este producto
Código de producto. 15561501
  • 3970.00€ / cada uno
Fecha de envío estimada
del proveedor 11-03-2019
Añadir a la cesta
Descripción
Descripción
Ambion™ siRNAs are classic 21-mers which incorporate the latest improvements in siRNA design, off-target effect prediction algorithms, and chemistry. These siRNAs are guaranteed-to-silence based on their proven design.
Especificación
Especificación
Ambion™ In-vivo Pre-designed siRNA, 1μmol, Solid, HPLC In-vivo Ready Purity, Combined Knockdown Effectiveness and Stability, Easy Tracking of Administered siRNAs, Double-Stranded siRNAs That are Recognized by the RNA-induced Silencing Complex (RISC) to Mediate Inhibition of a Target Gene
Ambion™ In-vivo Pre-designed siRNA
Room temperature
Solid
1μmol
HPLC In-vivo Ready
Videos
For Research Use Only. Not for use in diagnostic procedures.