Learn More
Alfa Aesar™ Aluminum trifluoromethanesulfonate
Marca: Alfa Aesar™ 038568.18
Detalles adicionales : CAS : 74974-61-1 Peso : 0.05000kg Transport : UN number : 3261 Chem class : 8 Pack group : III
Cantidad. | 50g |
---|
Identificadores de productos químicos
74974-61-1 | |
474.171 | |
FKOASGGZYSYPBI-UHFFFAOYSA-K | |
2737634 | |
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Al+3] |
C3AlF9O9S3 | |
MFCD00143596 | |
aluminum trifluoromethanesulfonate, aluminum triflate, aluminium triflate, aluminum tris trifluoromethanesulfonate, aluminium trifluoromethanesulphonate, aluminium 3+ tritriflate, aluminium trifluoromethanesulfonate, aluminum iii triflate, aluminium iii triflate, aluminum tris fluoranyl methanesulfonate | |
aluminum;trifluoromethanesulfonate |
Especificación
Aluminum trifluoromethanesulfonate | |
50g | |
~300°C | |
C3AlF9O9S3 | |
MFCD00143596 | |
aluminum trifluoromethanesulfonate, aluminum triflate, aluminium triflate, aluminum tris trifluoromethanesulfonate, aluminium trifluoromethanesulphonate, aluminium 3+ tritriflate, aluminium trifluoromethanesulfonate, aluminum iii triflate, aluminium iii triflate, aluminum tris fluoranyl methanesulfonate | |
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Al+3] | |
474.171 | |
474.2 |
74974-61-1 | |
White | |
Hygroscopic | |
Al(CF3SO3)3 | |
UN3261 | |
FKOASGGZYSYPBI-UHFFFAOYSA-K | |
aluminum;trifluoromethanesulfonate | |
2737634 | |
Powder |