missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Aluminum sulfate hydrate, Puratronic™, 99.999% (metals basis)

Marca:  Alfa Aesar™ 010629.14

Detalles adicionales : CAS : 17927-65-0 Peso : 0.02500kg

Cantidad. 25g

 Ver más versiones de este producto

Código de producto. 11310819

  • 161.00€ / 25 g


Stock EU 3
Añadir a la cesta

Especificaciones

Identificadores de productos químicos

17927-65-0
360.146
BUACSMWVFUNQET-UHFFFAOYSA-H
3080712
O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3]
Al2H2O13S3
MFCD00149136
aluminium sulfate hydrate, aluminum sulfate hydrate, dialuminum trisulfate hydrate, aluminum sulfate usp, aluminum sulfate 2:3 hydrate, alminum sulfate hydrate, aluminum sulfate usan, sulfuric acid, aluminum salt 3:2 , hydrate, 2al.3so4.h2o, aluminum sulfate hydrate
dialuminum;trisulfate;hydrate

Especificaciones

Aluminum sulfate hydrate
(metals basis)
MFCD00149136
aluminium sulfate hydrate, aluminum sulfate hydrate, dialuminum trisulfate hydrate, aluminum sulfate usp, aluminum sulfate 2:3 hydrate, alminum sulfate hydrate, aluminum sulfate usan, sulfuric acid, aluminum salt 3:2 , hydrate, 2al.3so4.h2o, aluminum sulfate hydrate
O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3]
360.146
342.15 (Anhydrous)
99.999%
1.61g/mL
3.5
17927-65-0
Al2H2O13S3
14,366
BUACSMWVFUNQET-UHFFFAOYSA-H
dialuminum;trisulfate;hydrate
3080712
Crystalline
Puratronic™
White
25g
Videos
SDS