Learn More
Brady™ Aluminum with Polyester Overlaminate Signs: Eating and Drinking Prohibited
Product Code. 11740294 Shop All Brady Products
Change view
Click to view available options
Diameter (Metric):
100 mm
200 mm
315 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
11740294 200 mm Each
11730294 100 mm Each
11724854 315 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11740294 Supplier Brady™ Supplier No. 241954
  • ONLINE EXCLUSIVE PRICE
    14.95€
    13.79€ / paquete de 1
    Ahorro 1.16€
    7% Descuento
Fecha estimada de envoi: 04-12-2026