Learn More
Brady™ Aluminum: ISO Safety Sign - Fire hose reel
Product Code. 15644377 Shop All Brady Products
Change view
Click to view available options
Width (Metric):
100 mm
200 mm
Height (Metric):
100 mm
200 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) Height (Metric) Unit Size
15644377 200 mm 200 mm Each
15634377 100 mm 100 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15644377 Supplier Brady™ Supplier No. 817053
  • ONLINE EXCLUSIVE PRICE
    11.50€
    11.28€ / paquete de 1
    Ahorro 0.22€
    1% Descuento
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Sign compliant to ISO 7010.
  • Aluminum
  • For indoor and outdoor use
Specifications
Certifications/Compliance ISO 7010
Alternate Part Number PIC F002-200x200-AL-CRD/1
Material Aluminum
Quantity 1 Ea.
For Use With (Application) Indoor and Outdoor
Height (Metric) 200 mm
Thickness 1.016 mm
Width (Metric) 200 mm
Temperature Range (Metric) -40°C to 140°C
Shape Square
Languages Global
Color White on Red