Learn More
Statebourne Cryogenics™ Aluminum Dewar Bio8 Liquid Nitrogen Storage
Marca:  Statebourne Cryogenics™ 9902050
Detalles adicionales : Peso : 0.80000kg
Código de producto. 11944215
  • 1291.29€ / cada uno
Consúltenos : 902 239 303 
Añadir a la cesta
Descripción
Descripción
  • Durable aluminium construction with super insulated high vacuum system to ensure high performance
  • Liquid nitrogen refrigerators with capacities from 2L to 36L offering a choice of holding times and specimen capacity
  • Lockable lids in high strength polycarbonate
  • CE marked under the Medical Devices Directive
  • Supplied with canisters and neckplug
Especificaciones
Especificaciones
8 L
Aluminum
Videos
Documentación Ver productos accesorios