Learn More
Statebourne Cryogenics™ Almacenamiento de nitrógeno líquido Cryolab en contenedores de aluminio
Supplied with neckplug and dipstick
en la oferta
Especificaciones
Material Aluminio
Ver más especificaciones
Descripción
Descripción
Comprehensive range of accessories available Series of liquid nitrogen dewars providing a range of capacities to suit individual laboratory storage and liquid handling requirements.
  • All aluminium construction
  • All units guaranteed for three years
  • Low pressure dispenser on models CL20 to 50
  • ISO 9001 designed and manufactured
  • Supplied with neckplugs and dipstick
Especificaciones
Especificaciones
Aluminio