missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole Parmer Alumina process tube, open both ends, 2in OD, 18in
Marca: Cole Parmer 33910-5
Alumina process tube, open both ends, 2in OD, 18in
Detalles adicionales : Peso : 0.95000kg