missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALS2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 13998-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The function of this protein remains unknown.Especificaciones
ALS2 | |
Polyclonal | |
Unconjugated | |
ALS2 | |
ALS2, ALS2CR6, Alsin, ALSJ, IAHSP, KIAA1563, PLSJ | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
363235, 57679, 74018 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P0C5Y8, Q920R0, Q96Q42 | |
Als2 | |
ALS2 Fusion Protein Ag5087 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |