Learn More
Ansell™ AlphaTec™MICROCHEM™4000 Model 212 Apron
Product Code. 32177083 Shop All Ansell Products
Change view
Click to view available options
Closure Type:
Waist Ties
Unit Size:
Pack of 20
Product Code. Closure Type Unit Size
32177083 Waist Ties Pack of 20
1 options
Product Code. 32177083 Supplier Ansell™ Supplier No. GR40W0021200
  • ONLINE EXCLUSIVE PRICE
    329.00€
    257.05€ / paquete de 20
    Ahorro 71.95€
    21% Descuento
Fecha estimada de envoi: 15-12-2026
Add to basket
Add to basket
MICROCHEM™ 4000 is designed to provide anexceptional barrier against organic and inorganicchemicals and biological agents
  • Tie fastening to waist
  • 100cm long tie fastening
TRUSTED_SUSTAINABILITY
Specifications
Type Apron
Size Universal
Material Multi-layer Barrier Laminate Fabric
Color Green
Disposable Single-use
Closure Type Waist Ties
Sterility Non-sterile
No. of Pockets None
Resistance Abrasions, acids, anti-static, bloodborne pathogens, chemicals
Certifications/Compliance EN 14605: 2005 +A1:2009, Type PB (3);EN 14605: 2005 +A1:2009, Type PB (4); EN14126: 2003, Types PB(3)-B, PB (4)-B; EN1149-1: 1995
Length (Metric) 100 cm
Style Alphatec 4000, Model 212
Seam Type Welded and Taped