Learn More
Ansell™ AlphaTec™ 3000 AVANT2 AIRline Ultrasonically Welded and Taped G05
Código de producto. 16835010
Click to view available options
Tamaño:
Small
Medium
Grande
X-Large
2X-Large
3X-Large
Tamaño de la unidad:
paquete de 4
Código de producto. 16835010
Marca: Ansell™ YE30T00075706
Solo quedan null
Añadir a la cesta
Añadir a la cesta
AlphaTec™ 3000 AVANT2 AIRline Ultrasonically welded and taped G05 is a fully encapsulated suit designed for use in combination with the permanently attached MICROCHEM™ AVANT STS V.1 airline regulator.
Fully encapsulated suit designed to be used with the permanently attached MICROCHEM™ AVANT STS V.1 airline regulator.
  • Features include air-channel ventilation system
  • Exceptional protection, over 200 chemicals permeation tested, including chemical warfare agents
  • Welded and taped seams, our highest barrier to liquids and particulates
  • Panoramic visor design ensures a good field of vision for the wearer
  • A range of sleeve options available, including double cuffs and permanently attached chemical protective gloves.
  • HEPA filter provides secondary protection from airline contamination and a SMC silencer ensures the noise inside the suit is always below 70 dB (even at maximum airflow)
  • Suit/Belt mounted regulator complete with a low flow warning whistle the AVANT STS regulator can be mounted on the suit and removed for reuse providing cleaning and decontamination is permissible.
  • Emergency rip cord permits rapid doffing of the suit in cases of emergency or undue distress to the wearer
  • Attached socks with elasticated boot overflap - Socks are designed to be worn inside chemical protective boots with the leg overflap worn outside to reduce the potential for chemical ingress
TRUSTED_SUSTAINABILITY
Especificaciones
Certificaciones/conformidad CE CATEGORY III (Type 3, EN 1073-1, EN 1149-5)
Color Yellow
Para utilizar con (aplicación) Opening and draining pumps valves or lines Filling, blending and charging of raw materials Contaminated Environments Chemical Handling Transferring liquids and solids Raw material sample collection Concentration mixing and palletisation Hazmat response Assembly and disassembly of large pipes Loading and unloading of proces equipment
Hood Type Totalmente encapsulado
Material Multi-layer Barrier Laminate Fabric
Tamaño 2X-Large
Estilo de tobillo Attached Socks with Elasticated Boot Overflap
Tipo de tapón Zipper
Estilo de manguito Double cuffs, finger and thumb loop
Desechables Reusable
Sexo Unisex
Estilo de cuello Hood
N.º de bolsillos None
Resistencia Chemical I Radioactive I Antistatic
Tipo de costura ULTRASONICALLY WELDED AND TAPED