Learn More
Novus Biologicals™ alpha-Synuclein Overexpression Lysate
Marca:  Novus Biologicals™ NBL1-16274
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18214490
  • 685.73€ / 0.10 miligramo
Fecha estimada de envío: 21-08-2024
para ver el stock
Descripción
Descripción
The alpha-Synuclein Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Especificaciones
Especificaciones
Western Blot
0.1 mg
Protein
-80°C. Avoid freeze-thaw cycles.
Native
The lysate was created in HEK293T cells, using plasmid ID RC2166 and based on accession number NM_345. The protein contains a C-terminal DDK tag.
RIPA buffer
Homo sapiens synuclein, alpha (non A4 component of amyloid precursor) (SNCA), transcript variant NACP140, mRNA.
Alzheimers Research, Apoptosis, Cell Biology, Cellular Markers, Neurodegeneration, Neuroscience