missing translation for 'onlineSavingsMsg'
Learn More

alpha-Galactosidase A/GLA Antibody (001), Alexa Fluor™ 750, Novus Biologicals™

Rabbit Monoclonal Antibody

Marca:  Bio-Techne NBP2-90047AF750

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213416

  • 605.82€ / 0.10 ml
Fecha estimada de envío: 14-08-2024
para ver el stock

Descripción

Descripción

alpha-Galactosidase A/GLA Monoclonal specifically detects alpha-Galactosidase A/GLA in Human samples. It is validated for ELISA.
Especificaciones

Especificaciones

alpha-Galactosidase A/GLA
Monoclonal
Alexa Fluor 750
50mM Sodium Borate
Rabbit
Protein A purified
RUO
2717
Store at 4°C in the dark.
IgG
ELISA
1
ELISA
Agalsidase, agalsidase alfa, Alpha-D-galactosidase A, Alpha-D-galactoside galactohydrolase, alpha-D-galactoside galactohydrolase 1, alpha-gal A, alpha-galactosidase A, EC 3.2.1, EC 3.2.1.22, GALA, galactosidase, alpha, melibiase
This antibody was obtained from a rabbit immunized with purified, recombinant Human alpha-Galactosidase A/GLA (AAP36507.1; Met 1-Leu 429).
0.1 mL
Primary
Human
Purified
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales