missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ alpha-D-Glucose-1-phosphate disodium salt tetrahydrate, 98%

Marca:  Alfa Aesar™ A12064.14

Detalles adicionales : CAS : 56401-20-8 Peso : 0.02500kg

Cantidad. 25g

 Ver más versiones de este producto

Código de producto. 11499952

  • 146.00€ / 25 g


En stock 1
Añadir a la cesta

Descripción

Descripción

Applications
alpha-D-Glucose-1-phosphate disodium salt tetra hydrate is a glucose molecule that can be used in cellular catabolism. It is used as an intermediate in chemical and pharmaceutical research.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed. Store in cool, dry conditions in well ventilated place.
Especificación

Identificadores de productos químicos

56401-20-8
306.114
AQMGMJDBUKKFOG-PKXGBZFFSA-N
131855210
C(C1C(C(C(C(O1)OP(=O)(O)O)O)O)O)O.[Na].[Na]
C6H13Na2O9P
MFCD00150839
alpha-d-glucose-1-phosphate disodium salt tetrahydrate
sodium;[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl] dihydrogen phosphate

Especificación

alpha-D-Glucose-1-phosphate disodium salt tetrahydrate
25g
C6H13Na2O9P
alpha-d-glucose-1-phosphate disodium salt tetrahydrate
C(C1C(C(C(C(O1)OP(=O)(O)O)O)O)O)O.[Na].[Na]
306.114
376.16 (304.10 Anhydrous)
~138°C
56401-20-8
MFCD00150839
AQMGMJDBUKKFOG-PKXGBZFFSA-N
sodium;[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl] dihydrogen phosphate
131855210
98%
Videos
SDS