Learn More
alpha-Chlorophenylacetyl Chloride, TRC
Product Code. 30349450
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Product Code. Quantity Unit Size
30349450 500 mg 500mg
1 options
This item is not returnable. View return policy
Product Code. 30349450 Supplier Toronto Research Chemicals Supplier No. TRCC376505500MG
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material alpha-Chlorophenylacetyl Chloride
CAS 2912-62-1
Molecular Formula C8 H6 Cl2 O
Quantity 500 mg
Synonym Chlorophenyl-acetyl Chloride, (±)-2-Chloro-2-phenylacetyl Chloride, 2-Chloro-2-phenylacetyl Chloride, 2-Phenyl-2-chloroacetyl Chloride, Chlorophenylacetyl Chloride, Phenyl(chloro)acetyl Chloride, α-Chloro-α-phenylacetyl Chloride, α-Chlorobenzeneacetyl Chloride, α-Chlorophenylacetyl Chloride
InChI Formula InChI=1S/C8H6Cl2O/c9-7(8(10)11)6-4-2-1-3-5-6/h1-5,7H
SMILES ClC(C(=O)Cl)c1ccccc1
IUPAC Name 2-chloro-2-phenylacetyl chloride
Molecular Weight (g/mol) 189.039
Formula Weight 187.98