missing translation for 'onlineSavingsMsg'
Learn More

alpha-Actinin 1 Antibody (1D10), DyLight 594, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Bio-Techne NBP2-59465DL594

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213210

  • 562.62€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

alpha-Actinin 1 Monoclonal antibody specifically detects alpha-Actinin 1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Especificaciones

Especificaciones

alpha-Actinin 1
Monoclonal
DyLight 594
actinin 1 smooth muscle, actinin, alpha 1, alpha-actinin 1, Alpha-actinin cytoskeletal isoform, alpha-actinin-1, F-actin cross-linking protein, FLJ40884, FLJ54432, Non-muscle alpha-actinin-1
Recombinant human alpha-Actinin 1 (1-249aa) purified from E. coli
0.1 mL
Angiogenesis, Cancer, Cell Biology, Cellular Markers
87
Store at 4°C in the dark.
IgG2a κ
Western Blot, Flow Cytometry, ELISA, Immunofluorescence
1D10
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales