missing translation for 'onlineSavingsMsg'
Learn More

Alpha 1 Fetoprotein Receptor Antibody (5E1), Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Bio-Techne NBP2-78787

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18214426

  • 341.24€ / 0.20 miligramo
Fecha estimada de envío: 14-08-2024
para ver el stock

Descripción

Descripción

Alpha 1 Fetoprotein Receptor Monoclonal specifically detects Alpha 1 Fetoprotein Receptor in Human samples. It is validated for Western Blot, Immunohistochemistry.
Especificaciones

Especificaciones

Alpha 1 Fetoprotein Receptor
Monoclonal
Unconjugated
PBS, pH 7.4 with 0.09% Sodium Azide
Mouse
Protein A purified
Primary
Human
Purified
Western Blot, Immunohistochemistry
50
Western Blot, Immunohistochemistry
Alpha-1-fetoprotein, Alpha-fetoglobulin, alpha-fetoprotein, FETA, HP, HPAFP
Full length native protein (purified) (Human).
0.2 mg
174
Store at 4°C. Do not freeze.
IgG2a
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales