missing translation for 'onlineSavingsMsg'
Learn More

Alil(clorometilo)dimetilsilano, +97 %

Catalog Number p-7051780
Click to view available options
Cantidad:
1 g
5 g
25 g
Este artículo no se puede devolver. Vea la política de devoluciones
75422-66-1
C6H13ClSi
148.71
MFCD00012045
UHHLOVCFFWGSMO-UHFFFAOYSA-N
allyl chloromethyl dimethylsilane, silane, chloromethyl dimethyl-2-propenyl, chloromethyl dimethylprop-2-en-1-ylsilane, acmc-1bfca, dimethylallylchloromethylsilane, 3-chloromethyldimethylsilyl-1-propene, allyl chloromethyl dimethyl dimethylsilane, silane,allyl, chloromethyl ,dimethyl, chloromethyl dimethyl prop-2-en-1-yl silane
556526
clorometil-dimetil-prop-2-enilsilano
C[Si](C)(CCl)CC=C
Este artículo no se puede devolver. Vea la política de devoluciones
75422-66-1
C6H13ClSi
148.71
MFCD00012045
UHHLOVCFFWGSMO-UHFFFAOYSA-N
allyl chloromethyl dimethylsilane, silane, chloromethyl dimethyl-2-propenyl, chloromethyl dimethylprop-2-en-1-ylsilane, acmc-1bfca, dimethylallylchloromethylsilane, 3-chloromethyldimethylsilyl-1-propene, allyl chloromethyl dimethyl dimethylsilane, silane,allyl, chloromethyl ,dimethyl, chloromethyl dimethyl prop-2-en-1-yl silane
556526
clorometil-dimetil-prop-2-enilsilano
C[Si](C)(CCl)CC=C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Punto de ebullición 165°C (50mmHg)
Nombre del producto químico o material Allyl(chloromethyl)dimethylsilane
Forma física Líquido
Cantidad 1 g
Número UN UN1993
Temperatura de inflamación 27°C (81°F)
Formula Weight (peso de la fórmula) 148.71
Porcentaje de pureza ≥97%
Índice de refracción 1.449
Sensibilidad Moisture sensitive
Densidad 0.907 g/mL
Mostrar más Mostrar menos
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'