Learn More
Aldosterone 21-Acetate, TRC
Product Code. 30326939
Change view
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.5mg
Product Code. Quantity Unit Size
30326939 2.5 mg 2.5mg
1 options
This item is not returnable. View return policy
Product Code. 30326939 Supplier Toronto Research Chemicals Supplier No. TRCA5147102.5MG
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Aldosterone 21-Acetate
CAS 2827-21-6
Molecular Formula C23H30O6
Quantity 2.5 mg
Synonym Aldosterone 21-Monoacetate, (11beta)-21-(Acetyloxy)-11,18-epoxy-18-hydroxy-pregn-4-ene-3,20-dione
InChI Formula InChI=1S/C23H30O6/c1-12(24)28-11-18(26)17-6-5-16-15-4-3-13-9-14(25)7-8-22(13,2)20(15)19-10-23(16,17)21(27)29-19/h9,15-17,19-21,27H,3-8,10-11H2,1-2H3/t15-,16-,17+,19-,20+,21?,22-,23+/m0/s1
SMILES CC(=O)OCC(=O)[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3[C@@H]5C[C@]12C(O)O5
IUPAC Name [2-[(1R,2S,5S,6S,14R,15S,16S)-18-hydroxy-14-methyl-11-oxo-17-oxapentacyclo[14.2.1.01,5.06,15.09,14]nonadec-9-en-2-yl]-2-oxoethyl] acetate
Molecular Weight (g/mol) 402.48
Formula Weight 402.2042