missing translation for 'onlineSavingsMsg'
Learn More

Aldolase C Antibody (2E11), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Bio-Techne NBP2-59408AF700

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18215570

  • 562.62€ / 0.10 ml
Fecha estimada de envío: 14-08-2024
para ver el stock

Descripción

Descripción

Aldolase C Monoclonal antibody specifically detects Aldolase C in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Especificaciones

Especificaciones

Aldolase C
Monoclonal
Alexa Fluor 700
ALDC, aldolase 3, aldolase C, fructose-bisphosphate, Brain-type aldolase, EC 4.1.2.13, fructoaldolase C, fructose-1,6-biphosphate triosephosphate lyase, fructose-bisphosphate aldolase C
Recombinant human Aldolase C (1-364aa) purified from E. coli
0.1 mL
Core ESC Like Genes, Stem Cell Markers
230
Store at 4°C in the dark.
IgG1 κ
Western Blot, Flow Cytometry, ELISA, Immunofluorescence
200000000000
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales