missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ALDOB (Human) Recombinant Protein
Human ALDOB full-length ORF ( AAH29399, 1 a.a. - 316 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00000229-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: MAHRFPALTQEQKKELSEIAQSIVANGKGILAADESVGTMGNRLQRIKVENTEENRRQFREILFSVDSSINQSIGGVILFHETLYQKDSQGKLFRNILKEKGIVVGIKLDQGGAPLAGTNKETTIQGLDGLSERCAQYKKDGVDFGKWRAVLRIADQCPSSLPIQENANALARYASICQQNGLVPIVEPEVIPDGDHDLEHCQYVTEKVLAAVYKALNDHHVYLEGTLLKPNMVTAGHACTKKYTPEQVAMATVTALHRTVPAAVPGICFLSGGMSEEDATLNLNAINLCPLPKPWKLSFSGVARLQTRRQPRRLL
Especificaciones
AAH29399 | |
aldolase B, fructose-bisphosphate | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
229 | |
Wheat germ expression system | |
10 μg | |
ALDOB | |
GST |