missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ALDH6A1 (Human) Recombinant Protein
Human ALDH6A1 full-length ORF ( AAH32371, 10 a.a. - 535 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00004329-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: VRARILQVSSKVKSSPTWYSASSFSSSVPTVKLFIGGKFVESKSDKWIDIHNPATNEVIGRVPQATKAEMDAAIASCKRAFPAWADTSVLSRQQVLLRYQQLIKENLKEIAKLITLEQGKTLADAEGDVFRGLQVVEHACSVTSLMMGETMPSITKDMDLYSYRLPLGVCAGIAPFNFPAMIPLWMFPMAMVCGNTFLMKPSERVPGATMLLAKLLQDSGAPDGTLNIIHGQHEAVNFICDHPDIKAISFVGSNKAGEYIFERGSRHGKRVQANMGAKNHGVVMPDANKENTLNQLVGAAFGAAGQRCMALSTAVLVGEAKKWLPELVEHAKNLRVNAGDQPGADLGPLITPQAKERVCNLIDSGTKEGASILLDGRKIKVKGYENGNFVGPTIISNVKPNMTCYKEEIFGPVLVVLETETLDEAIQIVNNNPYGNGTAIFTTNGATARKYAHLVDVGQVGVNVPIPVPLPMFSFTGSRSSFRGDTNFYGKQGIQFYTQLKTITSQWKEEDATLSSPAVVMPTMGR
Especificaciones
AAH32371.1 | |
aldehyde dehydrogenase 6 family, member A1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ALDH6A1 | |
GST |
4329 | |
Wheat germ expression system | |
25 μg | |
MGC40271, MMSADHA, MMSDH | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |