missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Albumin (Pig) ELISA Kit

Competitive Enzyme Immunoassay for the quantitative measurement of pig ALB in Plasma and Serum

Marca:  Abnova™ KA0497.1Kit

Detalles adicionales : Peso : 0.02000kg

 Ver más versiones de este producto

Código de producto. 16056135

  • 578.00€ / 1 set


Fecha estimada de envío
21-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

1.56 to 100ng/mL
Pig
1ng/mL
Quantification
Albumin (Pig) ELISA Kit
1 Kit
Cell Culture Supernatant
50μL
Biotin
Albumin (Pig) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of pig albumin.
Colorimetric
96 wells-antibody coated microwells, Swine albumin standard (3.2ug), Biotinylated Swine albumin antibody (1vial), MIX Diluent concentrate 10X (30mL), Wash Buffer Concentrate 20X (2 x 30mL), 80 Streptavidin-Peroxidase Conjugate (SP Conjugate), Chromogen Substrate (8mL), Stop Solution (12mL)
Albumin (Pig) ELISA kit
RUO
Store components of the kit at 4°C or -20°C as described in the protocol.
Videos
Documentos