missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Al-23 Tube, Both Ends Open, O.D.: 50mm, I.D.: 42mm, Alfa Aesar™
Marca: Alfa Aesar 031579.EO
Detalles adicionales : Peso : 0.01000kg
Especificación
Al-23 Tube | |
1000mm | |
Both ends open |