Learn More
Al-23 Fish Spine Beads;OD (mm), 3.5;Bore (mm), 1.0;Length (mm), 3.0
Product Code. 15496065
Change view
Click to view available options
Quantity:
100 Pk.
Unit Size:
Pack of 100
Product Code. Quantity Unit Size
15496065 100 Pk. Pack of 100
1 options
This item is not returnable. View return policy
Product Code. 15496065 Supplier Thermo Scientific Alfa Aesar Supplier No. 032905.KX
 Restricted Item
Fecha estimada de envoi: 17-12-2026
Add to basket
This item is not returnable. View return policy
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Al-23 Fish Spine Beads
Quantity 100 Pk.
Form Fish Spine Beads
What is the maximum working temperature of Al-23 Tubes?
The maximum working temperature for the Al-23 Tube is 1950 degrees C.