Learn More
AKT3 Antibody (66C1247), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Marca:  Novus Biologicals NB600-1298AF700
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18214435
  • 521.58€ / 0.10 ml
Fecha estimada de envío: 14-08-2024
para ver el stock
Descripción
Descripción
AKT3 Monoclonal specifically detects AKT3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Especificaciones
Especificaciones
AKT3
Monoclonal
Alexa Fluor 700
Q9Y243
AKT3
This monoclonal antibody was raised against a synthetic peptide corresponding to amino acid residues 119-136 (CSPTSQIDNIGEEEMDAS) of human Akt3, GenBank no. gi 4574744 gb AAD24196.1 AF135794_1. This sequence is identical in human, mouse, rat, cow, dog, and chicken.
0.1 mL
Cancer, Hypoxia, Phospho Specific, Protein Kinase, Signal Transduction
10000
Human, Mouse, Rat
Purified
Western Blot, Immunocytochemistry, Immunofluorescence
66C1247
Western Blot, Immunocytochemistry/Immunofluorescence
Akt-3, EC 2.7.11, EC 2.7.11.1, PKB gamma, PKBGDKFZp434N0250, PRKBG, v-akt murine thymoma viral oncogene homolog 3 (protein kinase B, gamma)
Mouse
Protein G purified
RUO
Primary
Cross-reacts with Human, Mouse and Rat. Not yet tested in other species.
Store at 4C in the dark.
IgG1