Learn More
Azlon™ Airtight Box
Product Code. 12084578
Change view
Click to view available options
Capacity (Metric):
500 mL
1000 mL
1500 mL
2000 mL
3500 mL
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
12084578 3500 mL Each
12035572 500 mL Each
12045572 1000 mL Each
12059632 1500 mL Each
12069632 2000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 12084578 Supplier Azlon Supplier No. BWY038
  • ONLINE EXCLUSIVE PRICE
    40.32€
    32.41€ / paquete de 1
    Ahorro 7.91€
    19% Descuento
View all promotions
Fecha estimada de envoi: 15-02-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Store a wide variety of substances in this versatile plastic box. Azlon™ Airtight Box is lightweight and unbreakable in normal use.
  • Made from tough HDPE
  • Tight fitting snap on lids create an airtight seal
  • Suitable for food contact applications in accordance with EU regulations
  • Suitable for general and refrigerated storage down to -50°C
Specifications
Product Type Airtight Box
Material HDPE
Includes Snap-on lid
For Use With (Equipment) For general and refrigerated storage down to –50°C
Dimensions (L x W x H) 210 x 210 x 65 mm
Capacity (Metric) 3500 mL
Height (Metric) 65 mm
Length (Metric) 210 mm
Width (Metric) 210 mm