Learn More
Aalborg™ Air Direct Glass Flow Tube 150mm Reference Scale
Product Code. 11798278
Change view
Click to view available options
Airflow Rate:
1.25 L/min.
10 L/min.
100 mL/min.
1093 mL/min.
143 mL/min.
1637 mL/min.
19 mL/min.
2.5 L/min.
200 mL/min.
23 L/min.
25 mL/min.
264 mL/min.
30 L/min.
300 mL/min.
374 mL/min.
42 L/min.
49 mL/min.
50 mL/min.
500 mL/min.
5188 mL/min.
70 mL/min.
75 mL/min.
8505 mL/min.
Unit Size:
Each
23 product options available for selection
Product selection table with 23 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Airflow Rate Unit Size
11798278 1093 mL/min. Each
11778308 75 mL/min. Each
11778348 264 mL/min. Each
11738348 49 mL/min. Each
11758328 2.5 L/min. Each
11788318 300 mL/min. Each
11768308 50 mL/min. Each
11718348 19 mL/min. Each
11778318 100 mL/min. Each
11748348 143 mL/min. Each
11778328 10 L/min. Each
11753199 25 mL/min. Each
11748368 70 mL/min. Each
11763199 200 mL/min. Each
11788258 374 mL/min. Each
11748328 1.25 L/min. Each
11773199 500 mL/min. Each
11788328 23 L/min. Each
11718368 1637 mL/min. Each
11708368 8505 mL/min. Each
11728288 5188 mL/min. Each
11733199 42 L/min. Each
11738288 30 L/min. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 23 options available.
23 options
This item is not returnable. View return policy
Product Code. 11798278 Supplier Aalborg™ Supplier No. FT08203SAVA
  • ONLINE EXCLUSIVE PRICE
    123.64€
    75.48€ / paquete de 1
    Ahorro 48.16€
    38% Descuento
 Restricted Item
Fecha estimada de envoi: 20-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Aalborg™ Air Direct Glass Flow Tube Reference Scale reults in superior mL and L readings by providing a scale of meaurements in a vairety of volumes.
  • Direct reading tubes are straightforward.
  • The measurement scale on each of these tubes reads actual flow at standard conditions in a choice of English or Metric units.
  • Obtaining actual flow rates with these tubes requires the use of a reference scale flow correlation table.
  • Reference scale tubes are useful when measuring flow rates for gases other than air and/or for non-standard conditions.
Specifications
Type Air Direct Flow Tube
For Use With (Application) For various fluids, gases or liquids, thanks to the correlation tables
Width (Metric) 150 mm
Airflow Rate 1093 mL/min.