missing translation for 'onlineSavingsMsg'
Learn More

AGTR1 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 2C5G11, Proteintech

Mouse Monoclonal Antibody

Marca:  Proteintech 66415-1-IG-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16818275

  • 140.00€ / 20 microlitros
Fecha estimada de envío: 17-04-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

AGTR1 is known as Angiotensin II Type 1 Receptor. Angiotensin II is a potent vasopressor hormone and a primary regulator of aldosterone secretion. It is an important effector controlling blood pressure and volume in the cardiovascular system. And it acts through at least two types of receptors. This gene encodes the type 1 receptor which is thought to mediate the major cardiovascular effects of angiotensin II. Moreover, this gene may play a role in the generation of reperfusion arrhythmias following restoration of blood flow to ischemic or infarcted myocardium. It was previously thought that a related gene, denoted as AGTR1B, existed; however, it is now believed that there is only one type 1 receptor gene in humans. Multiple alternatively spliced transcript variants have been reported for this gene.
Especificaciones

Especificaciones

AGTR1
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P25095, P29754, P30555, P30556
AGTR1, Agtr1a
AGTR1 Fusion Protein Ag14461
20 μL
Primary
Mouse, Porcine, Rat, Human
Antibody
IgG2a
Western Blot
2C5G11
Unconjugated
AGTR1
AG2S, AGTR1, AGTR1A, AGTR1B, Angiotensin II type 1 receptor, AT1, AT1AR, AT1B, AT1BR, AT1R, AT2R1, AT2R1A, AT2R1B, HAT1R, Type 1 angiotensin II receptor
Mouse
Protein A
RUO
100519976, 11607, 185, 24180
-20°C
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales