Learn More
Epredia™ Adcell™ PTFE Diagnostic Slides
Product Code. 17352650 Shop All Epredia Products
Change view
Click to view available options
Well Count:
3
12
Color:
Black
White
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Well Count Color Unit Size
17352650 3 Black Pack of 100
17342650 12 White Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17352650 Supplier Epredia™ Supplier No. X5XER203BAD1
  • ONLINE EXCLUSIVE PRICE
    134.00€
    124.99€ / paquete de 100
    Ahorro 9.01€
    6% Descuento
Fecha estimada de envoi: 04-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
These hydrophobic coatings bind liquids applied to the wells and keeps fluids in the well area. Clean, flat and suitable for immediate use
  • Binds and keeps liquids applied to the well area
  • PTFE Construction
  • Provides wettable wells for staining and growing cell cultures
  • Prevents cross-contamination
  • Hydrophobic coatings are very durable and withstand the strongest stains
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Length (Metric) 25 mm
Width (Metric) 75 mm
Material PTFE
Thickness (Metric) 1 mm
Type ADCELL
Well Count 3
Color Black
Diameter (Metric) 14 mm Well
Quantity 100/Pk.