missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15482-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene is a member of the ADAR (adenosine deaminase acting on RNA) family. Using site-specific adenosine modification, proteins encoded by these genes participate in the pre-mRNA editing of nuclear transcripts. The protein encoded by this gene, tRNA-specific adenosine deaminase 1, is responsible for the deamination of adenosine 37 to inosine in eukaryotic tRNA.Especificaciones
ADAT1 | |
Polyclonal | |
Unconjugated | |
ADAT1 | |
ADAT1, adenosine deaminase, tRNA 1, HADAT1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23536, 30947, 690810 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9BUB4, Q9JHI2 | |
Adat1 | |
ADAT1 Fusion Protein Ag7824 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |