Learn More
Interscience Adaptor for Petri Dish
Product Code. 13425199
Change view
Click to view available options
For Use With (Equipment):
Automatic colony counter, Scan 300/500/1200
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
13425199 Automatic colony counter, Scan 300/500/1200 Each
1 options
This item is not returnable. View return policy
Product Code. 13425199 Supplier Interscience Supplier No. 436005
  • ONLINE EXCLUSIVE PRICE
    139.00€
    137.38€ / paquete de 1
    Ahorro 1.62€
    1% Descuento
Fecha estimada de envoi: 21-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Adaptor for Petri dish (55 mm)
Adaptor for Petri dishes of dia. 55 mm for Scan 300/500/1200 (optional accessory)
Specifications
Description Adaptor for Petri dishes of dia. 55 mm for Scan 300/500/1200 (optional accessory)
Type Contact Petri Dish
For Use With (Equipment) Automatic colony counter, Scan 300/500/1200