missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACYP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12302-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Its physiological role is not yet clear.Especificaciones
ACYP2 | |
Polyclonal | |
Unconjugated | |
ACYP2 | |
Acylphosphatase 2, acylphosphatase 2, muscle type, ACYM, ACYP, ACYP2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
364224, 75572, 98 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P14621, P35745, P56375 | |
Acyp2 | |
ACYP2 Fusion Protein Ag2948 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |