missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACTL6A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10341-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a family member of actin-related proteins (ARPs), which share significant amino acid sequence identity to conventional actins. Both actins and ARPs have an actin fold, which is an ATP-binding cleft, as a common feature. The ARPs are involved in diverse cellular processes, including vesicular transport, spindle orientation, nuclear migration and chromatin remodeling. This gene encodes a 53 kDa subunit protein of the BAF (BRG1/brm-associated factor) complex in mammals, which is functionally related to SWI/SNF complex in S. cerevisiae and Drosophila; the latter is thought to facilitate transcriptional activation of specific genes by antagonizing chromatin-mediated transcriptional repression. Together with beta-actin, it is required for maximal ATPase activity of BRG1, and for the association of the BAF complex with chromatin/matrix. Three transcript variants that encode two different protein isoforms have been described.Especificaciones
ACTL6A | |
Polyclonal | |
Unconjugated | |
ACTL6A | |
actin like 6A, Actin like protein 6A, Actin related protein Baf53a, ACTL6, ACTL6A, Arp4, ARPN BETA, ArpNbeta, BAF53, BAF53A, BRG1 associated factor 53A, INO80 complex subunit K, INO80K | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
361925, 56456, 86 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O96019, Q9Z2N8 | |
Actl6a | |
ACTL6A Fusion Protein Ag0323 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |