missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbón activado, NORIT(R) SA 2, decolorante, Thermo Scientific Chemicals
Click to view available options
Cantidad:
500 g
2.5 kg
Envase:
Frasco de vidrio
Tambor de plástico
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ayuda de filtros
Especificaciones
Nombre del producto químico o material | Activated charcoal,NORIT* SA 2 |
Punto de fusión | 3652.0°C |
Color | Negro |
Pérdida por secado | 10% max. |
Envase | Tambor de plástico |
Cantidad | 2.5 kg |
Índice Merck | 15, 1807 |
Información de solubilidad | Solubility in water: insoluble. Other solubilities: insoluble in organic solvents,soluble matter in hno3: <=3%,soluble matter in alcohol: <=0.5% |
Formula Weight (peso de la fórmula) | 12 |
Forma física | Polvo |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'