missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACSM3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10168-2-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Has medium-chain fatty acid:CoA ligase activity with broad substrate specificity (in vitro). Acts on acids from C(4) to C(11) and on the corresponding 3-hydroxy- and 2,3- or 3,4-unsaturated acids (in vitro).Especificaciones
ACSM3 | |
Polyclonal | |
Unconjugated | |
ACSM3 | |
ACSM3, Butyrate CoA ligase 3, Protein SA homolog, SA, SAH | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20216, 24763, 6296 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q3UNX5, Q53FZ2, Q6SKG1 | |
ACSM3 | |
ACSM3 Fusion Protein Ag0224 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |