missing translation for 'onlineSavingsMsg'
Learn More

ACPP Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 15840-1-AP-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16815214

  • 109.00€ / 20 microlitros

Envío estimado: 29-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

ACPP is an enzyme which catalyzes the conversion of orthophosphoric monoester to alcohol and orthophosphate. It is synthesized under androgen regulation and is secreted by the epithelial cells of the prostate gland.
Especificaciones

Especificaciones

ACPP
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P15309, Q8CE08
acid phosphatase, prostate, ACP 3, ACP3, ACPP, PAP, Prostatic acid phosphatase
Rabbit
IgG
20 μL
-20°C
Primary
55, 56318
Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry
Unconjugated
ACPP
Liquid
ACPP
ACPP Fusion Protein Ag8592
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Videos
SDS
Documentos
Certificados