Learn More
Acetic Anhydride, Sigma-Aldrich™
Product Code. 15876730
Change view
Click to view available options
Quantity:
1 L
2.5 L
25 L
Unit Size:
1 L
2.5 L
25 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15876730 1 L 1 L
15851751 2.5 L 2.5 L
15861751 25 L 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
Product Code. 15876730 Supplier Sigma Aldrich Supplier No. 8222781000
 Restricted Item
Fecha estimada de envoi: 14-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
For synthesis
Acetic anhydride is a widely used reagent in organic synthesis. The reagent is effective for acetylating alcohols, amines, and thiols. Additionally, it is used in various named reactions such as the Pummerer reaction, the Perkin reaction, the Polonovski reaction, the N-oxide reaction, and the Thiele reaction. Acetic anhydride is an effective reagent during oxidation of alcohols, dehydration, ether cleavage, enol acetate formation, and gem-diacetate formation.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 108-24-7
Grade Research
Molecular Formula (CH3CO)2O
Molecular Weight (g/mol) 102.09
Specifications
Melting Point -73°C
Density 1.08 g/cm3 at 20°C
MDL Number MFCD00008705
Quantity 1 L
Synonym Acetic anhydride, Acetyl acetate, Acetyl oxide, Acetic anhydride
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Acetic anhydride for synthesis
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Acetic anhydride