Learn More
Aceclofenac, TRC
Product Code. 18043650
Change view
Click to view available options
Quantity:
10 mg
100 mg
1 g
Unit Size:
100mg
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18043650 100 mg 100mg
18021750 10 mg 10mg
18032350 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18043650 Supplier Toronto Research Chemicals Supplier No. TRCA130400100MG
 Restricted Item
Fecha estimada de envoi: 16-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 89796-99-6
Molecular Formula C16 H13 Cl2 N O4
Molecular Weight (g/mol) 354.18
Synonym Aceclofenac, Benzeneacetic acid, 2-[(2,6-dichlorophenyl)amino]-, carboxymethyl ester (9CI), 2-[[2-[(2,6-Dichlorophenyl)amino]phenyl]acetoxy]acetic acid, Aceclofar, Airtal, Biofenac, Bristaflam, Falcol, Gerbin, PR 82/3, Tresquim
IUPAC Name 2-[2-[2-(2,6-dichloroanilino)phenyl]acetyl]oxyacetic acid
SMILES OC(=O)COC(=O)Cc1ccccc1Nc2c(Cl)cccc2Cl
Specifications
Analyte or Component Names Aceclofenac
Shipping Condition Room Temperature
InChI Formula InChI=1S/C16H13Cl2NO4/c17-11-5-3-6-12(18)16(11)19-13-7-2-1-4-10(13)8-15(22)23-9-14(20)21/h1-7,19H,8-9H2,(H,20,21)
Solution Type Neat
Industry Type Pharmaceutical
Quantity 100 mg
Formula Weight 353.0222
Percent Purity >95%
Chemical Name or Material Aceclofenac
Grade HPLC