missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACE Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24743-1-AP-150UL
Additional Details : Peso : 0.00010kg
Beschreibung
This gene encodes an enzyme involved in catalyzing the conversion of angiotensin I into a physiologically active peptide angiotensin II. Angiotensin II is a potent vasopressor and aldosterone-stimulating peptide that controls blood pressure and fluid-electrolyte balance. This enzyme plays a key role in the renin-angiotensin system. Many studies have associated the presence or absence of a 287 bp Alu repeat element in this gene with the levels of circulating enzyme or cardiovascular pathophysiologies. Two most abundant alternatively spliced variants of this gene encode two isozymes - the somatic form and the testicular form that are equally active. Multiple additional alternatively spliced variants have been identified but their full length nature has not been determined.Spezifikation
ACE | |
Polyclonal | |
Unconjugated | |
ACE | |
ACE, ACE1, Angiotensin converting enzyme, CD143, DCP, DCP1, Dipeptidyl carboxypeptidase I, Kininase II, MVCD3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11421, 1636 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.36 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P09470, P12821 | |
Ace | |
ACE Fusion Protein Ag20654 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |