Learn More
ABT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca:  Proteintech 14148-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
 Ver más versiones de este producto
Código de producto. 16812664
  • 109.00€ / 20 microlitros
Envío estimado: 02-11-2021
Descripción
Descripción
Could be a novel TATA-binding protein (TBP) which can function as a basal transcription activator. Can act as a regulator of basal transcription for class II genes.
Especificaciones
Especificaciones
ABT1
0.47 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q4KLM5, Q9QYL7, Q9ULW3
ABT1, hABT1
Rabbit
IgG
20 μL
-20°C
Primary
29777, 306960, 30946
Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunocytochemistry
Unconjugated
ABT1
Liquid
ABT1
ABT1 Fusion Protein Ag5317
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat