missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ IgM (Human) ELISA Kit

Sandwich Enzyme Immunoassay for the quantitative measurement of Human IgM

418.00€


Includes:
  • Human IgM microplate: A 96-well polystyrene microplate (12 strips of 8 wells) coated with a polyclonal antibody against IgM, 96 (8 x 12) wells
  • Sealing tapes: Pressure-sensitive sealing tapes that can be cut to fit the format of the individual assay, 3 slides
  • Human IgM standard: Human IgM in a buffered protein base, lyophilized, 200ng
  • Biotinylated human IgM antibody (50X): A 50-fold concentrated biotinylated polyclonal antibody against human IgM, 140μL
  • Mix diluent concentrate (10X): A 10-fold concentrated buffered protein base, 30mL
  • Wash buffer concentrate (20X): A 20-fold concentrated buffered surfactant, 30mL x 2
  • Streptavidin-peroxidase conjugate (SP conjugate): A 100-fold concentrate, 80μL
  • Chromogen substrate: A ready-to-use stabilized peroxidase chromogen substrate tetramethylbenzidine, 8mL
  • Stop solution: A 0.5N hydrochloric acid to stop the chromogen substrate reaction, 12mL

Productos 1
Código de producto Marca Conjugado Precio Cantidad y disponibilidad  
Código de producto Marca Conjugado Precio Cantidad y disponibilidad  
16056885 Ver documentos Abnova™
KA1855.1KIT
Biotin 418.00€
1 set Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío
29-06-2020
Añadir a la cesta