missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ IgD (Human) ELISA Kit
highly sensitive two-site enzyme-linked immunoassay for the quantitative determination of IgD in Human biological samples.
en la oferta
Especificaciones
Método de detección | Colorimétrico |
---|---|
Tipo de producto | Kit de ELISA IgD (humana) |
Includes:
- Diluent Concentrate (Running Buffer): One bottle containing 5X concentrated diluent running buffer, 50mL
- Wash Solution Concentrate: One bottle containing 20X concentrated wash solution, 50mL
- Enzyme-Antibody Conjugate 100X: One vial containing affinity purified anti- human IgD antibody conjugated with horseradish peroxidase in stabilizing buffer, 150μL
- Chromogenic Substrate Solution: One vial containing 3,3',5,5'-tetramethybenzidine (TMB) and hydrogen peroxide in citric acid buffer at pH 3.3, 12mL
- Stop Solution: One vial containing 0.3M sulfuric acid (WARNING: Avoid contact with skin), 12mL
- Anti- human IgD ELISA micro plate: Twelve removable eight (8) well strips in well holder frame. Each well is coated with affinity-purified anti-human IgD, 96 wells
- Human IgD Calibrator: One vial containing human IgD Calibrator, 1 vial
Especificaciones
Colorimétrico | |
Kit de ELISA IgD (humana) |