Learn More
ABL1 [T315I] Recombinant Human Protein, Invitrogen™
ABL1 [T315I] Recombinant Human Protein
Marca:  Invitrogen PV3866
Detalles adicionales : Peso : 0.05250kg
Código de producto. 11533597
  • 632.00€ / 10 microgramos
Fecha estimada de envío: 16-04-2024
para ver el stock
Descripción
Descripción
The ABL1 proto-oncogene encodes a cytoplasmic and nuclear protein tyrosine kinase that has been implicated in processes of cell differentiation, cell division, cell adhesion, and stress response. The T315I mutant is a Gleevec resistant form of ABL1.
Especificaciones
Especificaciones
NP_005148.2, T315I
Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay
125.4kDa
10μg
For research use only. Not for use in diagnostic procedures
ABL1 T315I, ABL, c-ABL
ABL1 T315I was pre-diluted in enzyme dilution buffer and assayed in 50mM Tris (pH 7.5), 10mM MgCl2, 1mM DTT, 0.01% NP-40, 200μM ATP, 333μg/mL Abl1 peptide substrate (EAIYAAPFAKKK) and trace [32P]-γ-ATP for 10 minutes at 30°C
Full length (T315I)
104nmoles of phosphate transferred to Abl1 peptide substrate (EAIYAAPFAKKK) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 1.67μg/mL
His Tag
Kinase and inhibitors
60% by SDS-PAGE
0.39mg/mL
25
ABL1 [T315I] Recombinant Human Protein
–80°C
1 Each
ABL1 [T315I] Recombinant Human Protein
ABL
ABL1 [T315I] Recombinant Human Protein
Human
Baculovirus
Cytoplasmic tyrosine kinase
Sugerencias de productos
Sugerencias de productos
Videos
SDS
Documentos
Documentos
Certificados
Ofertas especiales
Ofertas especiales
For Research Use Only. Not for use in diagnostic procedures.