missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABL1 Recombinant Human Protein
ABL1 Recombinant Human Protein
Marca: Invitrogen PV3585
Detalles adicionales : Peso : 0.05250kg
Descripción
The ABL1 proto-oncogene encodes a cytoplasmic and nuclear protein tyrosine kinase that has been implicated in processes of cell differentiation, cell division, cell adhesion, and stress response.Especificaciones
NP_005148.2 | |
Protein Assays and Analysis, Kinase Assay | |
125.4kDa | |
1mg | |
For research use only. Not for use in diagnostic procedures | |
ABL1, ABL, c-ABL | |
ABL1 was pre-diluted in enzyme dilution buffer and assayed in 50mM Tris (pH 7.5), 10mM MgCl2, 1mM DTT, 0.01% NP-40, 200μM ATP, 100μM Abl1 peptide substrate (EAIYAAPFAKKK) and trace [32P]-γ-ATP for 10 minutes at 30°C | |
ABL1 Recombinant Human Protein | |
Human | |
Baculovirus | |
80% by SDS-PAGE |
0.2mg/mL | |
Frozen | |
ABL1 Recombinant Human Protein | |
–80°C | |
1 Each | |
ABL1 Recombinant Human Protein | |
ABL | |
2090nmoles of phosphate transferred to Abl1 peptide substrate (EAIYAAPFAKKK) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 0.4μg/mL | |
His Tag | |
Frozen |
For Research Use Only. Not for use in diagnostic procedures.