missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABI1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 27387-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a member of the DEAD box protein family. DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. The nucleolar protein encoded by this gene interacts in a hormone-dependent manner with nuclear receptors, and represses their transcriptional activity. Alternative splice variants that encode different isoforms have been found for this gene.Especificaciones
ABI1 | |
Polyclonal | |
Unconjugated | |
ABI1 | |
Abelson interactor 1, ABI 1, ABI1, Abl binding protein 4, abl interactor 1, AblBP4, E3B1, Eps8 binding protein, Nap1 binding protein, NAP1BP, SSH3BP, SSH3BP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10006 | |
-20°C | |
Liquid |
Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8IZP0 | |
ABI1 | |
ABI1 Fusion Protein Ag26537 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |