missing translation for 'onlineSavingsMsg'
Learn More

ABCB5 Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody

Marca:  Bio-Techne NBP1-77687UV

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213289

  • 561.54€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

ABCB5 Polyclonal specifically detects ABCB5 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Especificaciones

Especificaciones

ABCB5
Polyclonal
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
ABCB5 P-gp, ABCB5alpha, ABCB5beta, ATP-binding cassette protein, ATP-binding cassette sub-family B member 5, ATP-binding cassette, sub-family B (MDR/TAP), member 5, EC 3.6.3, EC 3.6.3.44, EST422562, P-glycoprotein ABCB5
Rabbit
0.1 mL
340273
Store at 4°C in the dark.
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
DyLight 350
50 mM sodium borate with 0.05% sodium azide
ABCB5
A synthetic peptide made to an internal portion of the human ABCB5 protein (between residues 150-200) [UniProt Q2M3G0]
Primary
Human, Mouse
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales